Assay Details
| Gene Symbol: Dda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001294259.1 | NP_001281188.1 | 2 - 3 | 182 | 66 | |
| NM_025600.2 | NP_079876.1 | 3 - 4 | 263 | 66 | ||
| NR_122115.1 | 3 - 4 | 259 | 66 | |||
| GenBank mRNA | AK006318.1 | - | 3 - 4 | 247 | 66 | |
| AK007085.1 | - | 3 - 4 | 282 | 66 | ||
| AK032966.1 | - | 3 - 4 | 221 | 66 | ||
| AK149980.1 | - | 3 - 4 | 243 | 66 | ||
| AK160132.1 | - | 3 - 4 | 260 | 66 | ||
| AK168085.1 | - | 3 - 4 | 221 | 66 | ||
| BB610194.1 | - | 3 - 4 | 256 | 66 | ||
| BC034701.1 | - | 3 - 4 | 223 | 3494430 | 66 | |
| BC093523.1 | - | 3 - 4 | 206 | 3711509 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dda1
Genomic Map