Assay Details
| Gene Symbol: Plvap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032398.2 | NP_115774.2 | 4 - 5 | 1277 | 90 | |
| GenBank mRNA | AF348403.1 | - | 4 - 5 | 1248 | 90 | |
| AF369900.1 | - | 4 - 5 | 1272 | 90 | ||
| BC013517.1 | - | 4 - 5 | 1280 | 4235957 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plvap
Genomic Map