Assay Details
Gene Symbol: Ocel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029865.2 | NP_084141.2 | 2 - 3 | 850 | 107 | |
GenBank mRNA | AK155219.1 | - | 2 - 3 | 810 | 107 | |
BC019407.1 | - | 2 - 3 | 806 | 5151818 | 107 | |
BY542600.1 | - | 1 - 2 | 122 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956545 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956551 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956548 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956549 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956547 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956546 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2057622 | AK143433.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2180869 | BC019407.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568440 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P136430 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2180869 | BC019407.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2057622 | AK143433.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956543 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956559 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956554 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956557 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956565 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956548 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956552 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956550 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956556 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956546 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956562 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25956549 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P136430 | - | - |
Whole Mouse Genome G4122A | A_51_P422030 | BC019407.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P136430 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06814834 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956546 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956551 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956547 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956549 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956548 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25956545 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956627 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956610 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956522 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956565 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956531 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956550 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956612 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956527 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956596 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956615 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956532 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956633 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956552 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956516 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956630 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956512 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956528 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956530 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956559 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956635 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956554 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956546 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956602 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956594 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956525 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956609 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956555 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956521 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956557 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956523 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956513 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956519 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956518 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956533 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956548 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25956606 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956552 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06814843 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956556 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956559 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956557 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956562 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956546 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956565 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07338524 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25956544 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10572592 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10572595 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10572594 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10572593 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1425191_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10572591 | - | - |
Mouse Genome 430 2.0 Array | 1425191_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1425191_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77090_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 77090_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800005629.mm.1 | NM_029865.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005629.mm.1 | NM_029865.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005629.mm.1 | TC0800000784.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800005623.mm.1 | NM_029865.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005623.mm.1 | NM_029865.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005623.mm.1 | TC0800000784.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800005628.mm.1 | NM_029865.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005628.mm.1 | NM_029865.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005628.mm.1 | TC0800000784.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800005626.mm.1 | NM_029865.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005626.mm.1 | NM_029865.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005626.mm.1 | TC0800000784.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800005632.mm.1 | NM_029865.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005632.mm.1 | NM_029865.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005632.mm.1 | TC0800000784.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0800005633.mm.1 | NM_029865.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005633.mm.1 | NM_029865.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800005633.mm.1 | TC0800000784.mm.1 | MTA TC |
Mouse Expression Set 430 | 1425191_at | - | - |
Mouse Expression Set 430 | 1425191_PM_at | - | - |
Murine Genome U74v2 Set | 104444_at | - | - |
Murine Genome U74v2 Set | 104444_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1425191_at | - | - |
Mouse Genome 430A 2.0 Array | 1425191_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425191_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1425191_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 920374 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2667463 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2667463 | - | - |
