Assay Details
| Gene Symbol: Pak1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026550.3 | NP_080826.2 | 4 - 5 | 762 | 102 | |
| GenBank mRNA | AF386076.1 | - | 4 - 5 | 748 | 102 | |
| AK002852.1 | - | 4 - 5 | 512 | 102 | ||
| AK077846.1 | - | 4 - 5 | 750 | 102 | ||
| AK077941.1 | - | 4 - 5 | 509 | 102 | ||
| AK135946.1 | - | 4 - 5 | 509 | 102 | ||
| AK150747.1 | - | 4 - 5 | 510 | 102 | ||
| AK150919.1 | - | 4 - 5 | 510 | 102 | ||
| AY030406.1 | - | 4 - 5 | 577 | 102 | ||
| BC018522.1 | - | 4 - 5 | 505 | 102 | ||
| BC043461.1 | - | 4 - 5 | 678 | 102 | ||
| BC043725.1 | - | 4 - 5 | 678 | 102 | ||
| BC116729.1 | - | 4 - 5 | 493 | 102 | ||
| BC116731.1 | - | 4 - 5 | 493 | 102 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1ip1
Genomic Map