Assay Details
| Gene Symbol: Pak1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026550.3 | NP_080826.2 | 6 - 7 | 960 | 69 | |
| GenBank mRNA | AF386076.1 | - | 6 - 7 | 946 | 69 | |
| AK002852.1 | - | 6 - 7 | 710 | 69 | ||
| AK077846.1 | - | 6 - 7 | 948 | 69 | ||
| AK077941.1 | - | 6 - 7 | 707 | 69 | ||
| AK135946.1 | - | 6 - 7 | 707 | 69 | ||
| AK150747.1 | - | 6 - 7 | 708 | 69 | ||
| AK150919.1 | - | 6 - 7 | 708 | 69 | ||
| AY030406.1 | - | 6 - 7 | 775 | 69 | ||
| BC018522.1 | - | 6 - 7 | 703 | 69 | ||
| BC043461.1 | - | 6 - 7 | 876 | 69 | ||
| BC043725.1 | - | 6 - 7 | 876 | 69 | ||
| BC060371.1 | - | 5 - 6 | 984 | 69 | ||
| BC116729.1 | - | 6 - 7 | 691 | 69 | ||
| BC116731.1 | - | 6 - 7 | 691 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pak1ip1
Genomic Map