Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800017193.mm.1 |
NM_001167981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017193.mm.1 |
NM_001167981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017193.mm.1 |
TC0800002256.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800017194.mm.1 |
NM_001167981.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017194.mm.1 |
NM_001167981.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017194.mm.1 |
NM_144731.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017194.mm.1 |
NM_144731.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800017194.mm.1 |
TC0800002256.mm.1 |
MTA TC |