Assay Details
| Gene Symbol: Mef2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170537.1 | NP_001164008.1 | 9 - 10 | 1514 | 71 | |
| NM_025282.3 | NP_079558.1 | 9 - 10 | 1508 | 71 | ||
| GenBank mRNA | AK009139.1 | - | 9 - 10 | 1348 | 71 | |
| AK077603.1 | - | 8 - 9 | 1307 | 71 | ||
| AK132573.1 | - | 9 - 10 | 1343 | 71 | ||
| BC026841.1 | - | 9 - 10 | 1270 | 4481227 | 71 | |
| BC037731.1 | - | 9 - 10 | 1260 | 4500786 | 71 | |
| BC057650.1 | - | 9 - 10 | 1249 | 4500786 | 71 | |
| L13171.1 | - | 7 - 8 | 1075 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mef2c
Genomic Map