Assay Details
| Gene Symbol: Gla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013463.2 | NP_038491.2 | 2 - 3 | 448 | 80 | |
| GenBank mRNA | AK040081.1 | - | 2 - 3 | 380 | 80 | |
| AK054547.1 | - | 2 - 3 | 400 | 80 | ||
| AK145011.1 | - | 2 - 3 | 390 | 80 | ||
| AK145148.1 | - | 448 | 80 | |||
| AK164231.1 | - | 2 - 3 | 381 | 80 | ||
| AK166204.1 | - | 2 - 3 | 381 | 80 | ||
| AK166245.1 | - | 2 - 3 | 397 | 80 | ||
| BC009021.1 | - | 2 - 3 | 401 | 3708950 | 80 | |
| U34071.1 | - | 2 - 3 | 398 | 80 | ||
| U50716.1 | - | 2 - 3 | 386 | 80 | ||
More Information
Set Membership: |
|

Gla
Genomic Map