Assay Details
| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043344.1 | - | 19 - 20 | 2073 | 76 | |
| AK145200.1 | - | 20 - 21 | 2129 | 76 | ||
| AK161473.1 | - | 20 - 21 | 2212 | 76 | ||
| BC024848.1 | - | 3 - 4 | 725 | 76 | ||
| BC030328.1 | - | 20 - 21 | 2129 | 5376020 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cenpi
Genomic Map