Assay Details
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030174.2 | NP_084450.2 | 14 - 15 | 1747 | 125 | |
GenBank mRNA | AK047562.1 | - | 14 - 15 | 1748 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
