Assay Details
| Gene Symbol: Slc38a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172479.3 | NP_766067.2 | 14 - 15 | 1338 | 74 | |
| GenBank mRNA | AK054485.1 | - | 14 - 15 | 1325 | 74 | |
| AK077667.1 | - | 14 - 15 | 1307 | 74 | ||
| AK155928.1 | - | 14 - 15 | 1291 | 74 | ||
| AK158125.1 | - | 13 - 14 | 1459 | 74 | ||
| BC058856.1 | - | 5 - 6 | 596 | 74 | ||
| BC152401.1 | - | 14 - 15 | 1307 | 40057282 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc38a5
Genomic Map