Assay Details
| Gene Symbol: Slc38a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172479.3 | NP_766067.2 | 1 - 2 | 125 | 88 | |
| GenBank mRNA | AK054485.1 | - | 1 - 2 | 112 | 88 | |
| AK077667.1 | - | 1 - 2 | 94 | 88 | ||
| AK155928.1 | - | 1 - 2 | 78 | 88 | ||
| BC152401.1 | - | 1 - 2 | 94 | 40057282 | 88 | |
| BY247469.1 | - | 1 - 2 | 115 | 88 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077667.1, AK155928.1, BC152401.1, NM_172479.3 transcripts.
Set Membership: |
|

Slc38a5
Genomic Map