Assay Details
| Gene Symbol: Tfe3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105196.1 | NP_001098666.1 | 1 - 2 | 403 | 104 | |
| NM_001271489.1 | NP_001258418.1 | 1 - 2 | 403 | 104 | ||
| NM_172472.3 | NP_766060.2 | 1 - 2 | 403 | 104 | ||
| GenBank mRNA | AF510653.1 | - | 1 - 2 | 245 | 104 | |
| AK139756.1 | - | 1 - 2 | 400 | 104 | ||
| AK144657.1 | - | 1 - 2 | 368 | 104 | ||
| AK144751.1 | - | 1 - 2 | 389 | 104 | ||
| AK145832.1 | - | 1 - 2 | 387 | 104 | ||
| AK150372.1 | - | 1 - 2 | 156 | 104 | ||
| AK153051.1 | - | 1 - 2 | 156 | 104 | ||
| AK154690.1 | - | 1 - 2 | 369 | 104 | ||
| AK159162.1 | - | 1 - 2 | 303 | 104 | ||
| BC032963.1 | - | 1 - 2 | 284 | 104 | ||
| BC051568.1 | - | 1 - 2 | 356 | 104 | ||
| BC056358.1 | - | 1 - 2 | 391 | 6813486 | 104 | |
| BC063047.1 | - | 1 - 2 | 391 | 6813486 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001271489.1 transcripts.
Set Membership: |
|

Tfe3
Genomic Map