Assay Details
| Gene Symbol: Kif15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010620.1 | NP_034750.1 | 8 - 9 | 903 | 59 | |
| GenBank mRNA | AJ560623.1 | - | 8 - 9 | 920 | 59 | |
| AK052362.1 | - | 3 - 4 | 326 | 59 | ||
| BC060710.1 | - | 8 - 9 | 903 | 6409082 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kif15
Genomic Map