Assay Details
Gene Symbol: Gm6938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033482.1 | 2 - 3 | 253 | 83 | ||
GenBank mRNA | BE948818.1 | - | 1 - 2 | 83 | ||
BF470644.1 | - | 2 - 3 | 253 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
