Assay Details
Gene Symbol: Glipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027450.3 | NP_081726.1 | 1 - 2 | 74 | 70 | |
GenBank mRNA | AK089960.1 | - | 1 - 2 | 54 | 70 | |
AK141459.1 | - | 1 - 2 | 62 | 70 | ||
AK154433.1 | - | 1 - 2 | 52 | 70 | ||
AK167595.1 | - | 1 - 2 | 74 | 70 | ||
BC031750.1 | - | 1 - 2 | 74 | 4037002 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK089960.1, AK154433.1, AK167595.1, BC031750.1, NM_027450.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
