Assay Details
Gene Symbol: Glipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027450.3 | NP_081726.1 | 2 - 3 | 178 | 90 | |
GenBank mRNA | AK017557.1 | - | 2 - 3 | 155 | 90 | |
AK154433.1 | - | 2 - 3 | 156 | 90 | ||
AK167595.1 | - | 2 - 3 | 178 | 90 | ||
BC031750.1 | - | 2 - 3 | 178 | 4037002 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
