Assay Details
Gene Symbol: Prol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008644.2 | NP_032670.2 | 1 - 2 | 89 | 82 | |
GenBank mRNA | AF247816.1 | - | 1 - 2 | 91 | 82 | |
BC021158.1 | - | 1 - 2 | 93 | 82 | ||
BC021401.1 | - | 1 - 2 | 91 | 4166946 | 82 | |
BC042902.1 | - | 1 - 2 | 87 | 4924369 | 82 | |
U37531.1 | - | 1 - 2 | 89 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF247816.1, BC021401.1, BC042902.1, NM_008644.2, U37531.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
