Assay Details
| Gene Symbol: Padi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008812.2 | NP_032838.2 | 8 - 9 | 1033 | 106 | |
| GenBank mRNA | AK148156.1 | - | 8 - 9 | 1020 | 106 | |
| AK154814.1 | - | 8 - 9 | 1021 | 106 | ||
| AK158382.1 | - | 8 - 9 | 1020 | 106 | ||
| AK168685.1 | - | 5 - 6 | 742 | 106 | ||
| AK171275.1 | - | 8 - 9 | 1020 | 106 | ||
| BC040350.1 | - | 8 - 9 | 975 | 4192742 | 106 | |
| BC049947.1 | - | 8 - 9 | 1006 | 3600978 | 106 | |
| D16580.1 | - | 8 - 9 | 966 | 106 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Padi2
Genomic Map