Assay Details
| Gene Symbol: Padi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008812.2 | NP_032838.2 | 9 - 10 | 1147 | 95 | |
| GenBank mRNA | AK033760.1 | - | 1 - 2 | 120 | 95 | |
| AK154814.1 | - | 9 - 10 | 1135 | 95 | ||
| AK158382.1 | - | 9 - 10 | 1134 | 95 | ||
| AK168685.1 | - | 6 - 7 | 856 | 95 | ||
More Information
Set Membership: |
|

Padi2
Genomic Map