Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400031792.mm.1 |
TC0400003856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031792.mm.1 |
NM_198610.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031792.mm.1 |
NM_198610.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031793.mm.1 |
TC0400003856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031793.mm.1 |
NM_198610.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031793.mm.1 |
NM_198610.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031794.mm.1 |
TC0400003856.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400031794.mm.1 |
NM_198610.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400031794.mm.1 |
NM_198610.2 |
Refseq |