Assay Details
Gene Symbol: Cdkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276315.1 | NP_001263244.1 | 5 - 6 | 1193 | 120 | |
NM_016912.2 | NP_058608.1 | 5 - 6 | 927 | 120 | ||
NM_177270.4 | NP_796244.2 | 5 - 6 | 927 | 120 | ||
GenBank mRNA | AB029065.1 | - | 5 - 6 | 903 | 120 | |
AB029066.1 | - | 5 - 6 | 891 | 120 | ||
AB029067.1 | - | 5 - 6 | 1181 | 120 | ||
AK030598.1 | - | 5 - 6 | 1182 | 120 | ||
AK139820.1 | - | 5 - 6 | 1135 | 120 | ||
AK144574.1 | - | 5 - 6 | 928 | 120 | ||
AK145688.1 | - | 5 - 6 | 1076 | 120 | ||
BC117894.1 | - | 5 - 6 | 836 | 40096852 | 120 | |
BC117895.1 | - | 5 - 6 | 836 | 40096853 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
