Assay Details
Gene Symbol: Cdkl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276315.1 | NP_001263244.1 | 7 - 8 | 1431 | 149 | |
NM_016912.2 | NP_058608.1 | 7 - 8 | 1165 | 149 | ||
NM_177270.4 | NP_796244.2 | 7 - 8 | 1165 | 149 | ||
GenBank mRNA | AB029065.1 | - | 7 - 8 | 1141 | 149 | |
AB029066.1 | - | 7 - 8 | 1129 | 149 | ||
AK030598.1 | - | 7 - 8 | 1420 | 149 | ||
AK139820.1 | - | 7 - 8 | 1373 | 149 | ||
AK144574.1 | - | 7 - 8 | 1166 | 149 | ||
AK145688.1 | - | 7 - 8 | 1314 | 149 | ||
BC117894.1 | - | 7 - 8 | 1074 | 40096852 | 149 | |
BC117895.1 | - | 7 - 8 | 1074 | 40096853 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
