Assay Details
| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030235.1 | NP_084511.1 | 10 - 11 | 1427 | 96 | |
| GenBank mRNA | AK020003.1 | - | 10 - 11 | 1404 | 96 | |
| AK083605.1 | - | 10 - 11 | 1384 | 96 | ||
| AK122228.1 | - | 10 - 11 | 1405 | 96 | ||
| AK147365.1 | - | 10 - 11 | 1429 | 96 | ||
| BC117878.1 | - | 10 - 11 | 1389 | 40095961 | 96 | |
| BC117879.1 | - | 10 - 11 | 1389 | 96 | ||
More Information
Set Membership: |
|

Avl9
Genomic Map