Assay Details
| Gene Symbol: Avl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030235.1 | NP_084511.1 | 6 - 7 | 744 | 118 | |
| GenBank mRNA | AK020003.1 | - | 6 - 7 | 721 | 118 | |
| AK083605.1 | - | 6 - 7 | 700 | 118 | ||
| AK122228.1 | - | 6 - 7 | 722 | 118 | ||
| AK147365.1 | - | 6 - 7 | 746 | 118 | ||
| BC117878.1 | - | 6 - 7 | 706 | 40095961 | 118 | |
| BC117879.1 | - | 6 - 7 | 706 | 118 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Avl9
Genomic Map