Assay Details
| Gene Symbol: Vopp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146168.1 | NP_666280.1 | 1 - 2 | 194 | 143 | |
| GenBank mRNA | AK030975.1 | - | 1 - 2 | 181 | 143 | |
| AK031812.1 | - | 1 - 2 | 199 | 142 | ||
| AK158041.1 | - | 206 | 143 | |||
| AK165256.1 | - | 1 - 2 | 223 | 143 | ||
| AK170152.1 | - | 1 - 2 | 145 | 143 | ||
| AK170832.1 | - | 1 - 2 | 145 | 143 | ||
| BC024822.1 | - | 1 - 2 | 194 | 5359195 | 143 | |
| BC024884.1 | - | 1 - 2 | 115 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK030975.1, AK031812.1, AK158041.1, AK170152.1, AK170832.1, BC024822.1, NM_146168.1 transcripts.
Set Membership: |
|

Vopp1
Genomic Map