Assay Details
| Gene Symbol: Carhsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025821.2 | NP_080097.1 | 1 - 2 | 43 | 59 | |
| GenBank mRNA | AK002853.1 | - | 1 - 2 | 44 | 59 | |
| AK003926.1 | - | 1 - 2 | 46 | 59 | ||
| AK004711.2 | - | 1 - 2 | 43 | 59 | ||
| AK143612.1 | - | 1 - 2 | 44 | 59 | ||
| AK149875.1 | - | 1 - 2 | 47 | 59 | ||
| AK154460.1 | - | 1 - 2 | 44 | 59 | ||
| AK155318.1 | - | 1 - 2 | 46 | 59 | ||
| AK158112.1 | - | 1 - 2 | 46 | 59 | ||
| AK158252.1 | - | 1 - 2 | 46 | 59 | ||
| AK165373.1 | - | 1 - 2 | 46 | 59 | ||
| AK168504.1 | - | 1 - 2 | 44 | 59 | ||
| AK170411.1 | - | 1 - 2 | 41 | 59 | ||
| AK171282.1 | - | 1 - 2 | 46 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002853.1, AK003926.1, AK004711.2, AK143612.1, AK149875.1, AK154460.1, AK155318.1, AK158252.1, AK165373.1, AK168504.1, AK170411.1, AK171282.1, NM_025821.2 transcripts.
Set Membership: |
|

Carhsp1
Genomic Map