Assay Details
| Gene Symbol: Spen | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019763.2 | NP_062737.2 | 5 - 6 | 1598 | 70 | |
| GenBank mRNA | AB055980.1 | - | 4 - 5 | 1040 | 70 | |
| AF156529.1 | - | 4 - 5 | 1100 | 70 | ||
| AK137647.1 | - | 5 - 6 | 1486 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spen
Genomic Map