Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600000760.mm.1 |
NM_029253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000760.mm.1 |
NM_029253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000760.mm.1 |
NM_153123.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000760.mm.1 |
NM_153123.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000760.mm.1 |
TC1600000110.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000757.mm.1 |
NM_029253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000757.mm.1 |
NM_029253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000757.mm.1 |
NM_153123.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000757.mm.1 |
NM_153123.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000757.mm.1 |
TC1600000110.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600000761.mm.1 |
NM_029253.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000761.mm.1 |
NM_029253.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000761.mm.1 |
NM_153123.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000761.mm.1 |
NM_153123.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600000761.mm.1 |
TC1600000110.mm.1 |
MTA TC |