Assay Details
| Gene Symbol: Brd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK142152.1 | - | 4 - 5 | 519 | 100 | |
| BC031484.1 | - | 4 - 5 | 514 | 2654265 | 100 | |
| BC047372.1 | - | 4 - 5 | 498 | 100 | ||
| BC089379.1 | - | 4 - 5 | 472 | 100 | ||
More Information
Set Membership: |
|

Brd9
Genomic Map