Assay Details
| Gene Symbol: Cldn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016674.4 | NP_057883.1 | 1 - 2 | 442 | 76 | |
| GenBank mRNA | AF072127.1 | - | 1 - 2 | 384 | 76 | |
| AK028428.1 | - | 1 - 2 | 440 | 76 | ||
| AK036762.1 | - | 1 - 2 | 414 | 76 | ||
| AK036780.1 | - | 1 - 2 | 443 | 76 | ||
| AK040604.1 | - | 1 - 2 | 443 | 76 | ||
| AK054207.1 | - | 1 - 2 | 387 | 76 | ||
| AK081601.1 | - | 1 - 2 | 440 | 76 | ||
| EU076705.1 | - | 1 - 2 | 229 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cldn1
Genomic Map