Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
NM_001159317.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
NM_001159317.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
NM_001159318.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
NM_001159318.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
NM_008364.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
NM_008364.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
NM_134103.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
NM_134103.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003573.mm.1 |
TC1600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003574.mm.1 |
NM_134103.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003574.mm.1 |
NM_134103.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003574.mm.1 |
TC1600000376.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1600003575.mm.1 |
NM_134103.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003575.mm.1 |
NM_134103.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1600003575.mm.1 |
TC1600000376.mm.1 |
MTA TC |