Assay Details
| Gene Symbol: Cyp4f40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101588.1 | NP_001095058.1 | 11 - 12 | 1400 | 91 | |
| GenBank mRNA | BC158089.1 | - | 11 - 12 | 1477 | 9088175 | 91 |
More Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
menaquinone catabolic process
phylloquinone catabolic process
vitamin K catabolic process
oxidation-reduction process
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
menaquinone catabolic process
phylloquinone catabolic process
vitamin K catabolic process
oxidation-reduction process
oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, NAD(P)H as one donor, and incorporation of one atom of oxygen
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089500 | NM_001101588.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2089500 | BC158089.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 96249 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 842300 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491667 | AK016576.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491667 | AY013809.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491667 | BC066823.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 491667 | NM_033579.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783953 | AK139013.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783953 | BC026499.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783953 | BC050254.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783953 | BC145921.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783953 | BC145923.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 454345 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477635 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 82746 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174289 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478694 | AK042112.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478694 | XM_001473602.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478694 | XM_001473621.1 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 918176 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50258 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 110094 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089500 | NM_001101588.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2089500 | BC158089.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146412 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146486 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146532 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146457 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146508 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146452 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146432 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146561 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146478 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146445 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146465 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146475 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146409 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146417 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146407 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146377 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146403 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146448 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146494 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146524 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31146435 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146426 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146402 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13265897 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13358875 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13551083 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146498 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146412 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146472 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13551087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146414 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146392 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146459 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146439 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146451 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146380 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146434 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13358881 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146445 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13445978 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146449 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146475 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146477 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13283742 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146452 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146383 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146378 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13562966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146494 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13358878 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13079039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13551088 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13265893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13471396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146382 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146442 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13108404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13471384 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146376 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146448 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146450 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146502 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146508 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146431 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13471385 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146432 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146417 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13551085 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146446 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13194304 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13562964 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146429 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146461 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13471394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13235566 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146444 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146503 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13453527 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146447 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146379 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146399 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13079040 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13283744 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146505 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31146454 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10443887 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443896 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443894 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443895 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443889 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443888 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443897 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443892 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443891 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443893 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10443886 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10443885 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 885645 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1213968 | - | - |

Cyp4f40
Genomic Map