| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700005156.mm.1 |
NM_001101588.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005156.mm.1 |
NM_001101588.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005156.mm.1 |
TC1700000566.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700005154.mm.1 |
NM_001101588.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005154.mm.1 |
NM_001101588.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005154.mm.1 |
TC1700000566.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700005153.mm.1 |
NM_001101588.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005153.mm.1 |
NM_001101588.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005153.mm.1 |
TC1700000566.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700005155.mm.1 |
NM_001101588.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005155.mm.1 |
NM_001101588.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700005155.mm.1 |
TC1700000566.mm.1 |
MTA TC |