Assay Details
| Gene Symbol: Scn10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205321.1 | NP_001192250.1 | 9 - 10 | 1161 | 112 | |
| NM_009134.3 | NP_033160.2 | 9 - 10 | 1161 | 112 | ||
| GenBank mRNA | AK135787.1 | - | 6 - 7 | 782 | 112 | |
| AY538273.1 | - | 8 - 9 | 1091 | 112 | ||
More Information
Set Membership: |
|

Scn10a
Genomic Map