Assay Details
| Gene Symbol: Ndufb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019435.4 | NP_062308.2 | 2 - 3 | 636 | 89 | |
| GenBank mRNA | AK028121.1 | - | 2 - 3 | 357 | 89 | |
| BC027265.1 | - | 2 - 3 | 629 | 3589704 | 89 | |
| BC096047.1 | - | 2 - 3 | 332 | 89 | ||
| Y08702.1 | - | 2 - 3 | 329 | 89 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndufb11
Genomic Map