Assay Details
| Gene Symbol: Fgd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF402611.1 | - | 14 - 15 | 2086 | 91 | |
| AK028302.1 | - | 11 - 12 | 1535 | 91 | ||
| AK149626.1 | - | 14 - 15 | 2137 | 91 | ||
| BC129902.1 | - | 14 - 15 | 2173 | 40111637 | 91 | |
| BC129903.1 | - | 14 - 15 | 2174 | 40111639 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgd4
Genomic Map