Assay Details
| Gene Symbol: Dnm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025947.2 | NP_001021118.1 | 11 - 12 | 1521 | 69 | |
| NM_001276340.1 | NP_001263269.1 | 11 - 12 | 1539 | 69 | ||
| NM_001276341.1 | NP_001263270.1 | 10 - 11 | 1283 | 69 | ||
| NM_152816.3 | NP_690029.2 | 12 - 13 | 1560 | 69 | ||
| NR_075074.1 | 11 - 12 | 1330 | 69 | |||
| GenBank mRNA | AB079133.1 | - | 11 - 12 | 1373 | 69 | |
| AK051443.1 | - | 11 - 12 | 1450 | 69 | ||
| AK080871.1 | - | 11 - 12 | 1441 | 69 | ||
| AK154123.1 | - | 11 - 12 | 1290 | 69 | ||
| AK190489.1 | - | 1 - 2 | 101 | 69 | ||
| BC040777.1 | - | 10 - 11 | 1253 | 4948368 | 69 | |
| BC079635.1 | - | 11 - 12 | 1370 | 6400313 | 69 | |
| BC085158.1 | - | 12 - 13 | 1465 | 69 | ||
| BQ550714.1 | - | 3 - 4 | 457 | 69 | ||
| CD559921.1 | - | 3 - 4 | 383 | 69 | ||
| CK391623.1 | - | 3 - 4 | 308 | 69 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Dnm1l
Genomic Map