Assay Details
| Gene Symbol: Cd96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032465.2 | NP_115854.2 | 13 - 14 | 1728 | 85 | |
| GenBank mRNA | AK040539.1 | - | 13 - 14 | 1732 | 85 | |
| AK156470.1 | - | 13 - 14 | 1729 | 85 | ||
| AY029156.1 | - | 13 - 14 | 1586 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd96
Genomic Map