Assay Details
| Gene Symbol: Cd96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032465.2 | NP_115854.2 | 5 - 6 | 950 | 84 | |
| GenBank mRNA | AK040329.1 | - | 5 - 6 | 837 | 84 | |
| AK040539.1 | - | 5 - 6 | 954 | 84 | ||
| AK154076.1 | - | 5 - 6 | 920 | 84 | ||
| AK156470.1 | - | 5 - 6 | 951 | 84 | ||
| AY029156.1 | - | 5 - 6 | 811 | 84 | ||
| BC052865.1 | - | 5 - 6 | 847 | 30072864 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cd96
Genomic Map