Assay Details
| Gene Symbol: Cd96 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032465.2 | NP_115854.2 | 8 - 9 | 1315 | 120 | |
| GenBank mRNA | AK040539.1 | - | 8 - 9 | 1319 | 120 | |
| AK154076.1 | - | 8 - 9 | 1285 | 120 | ||
| AK156470.1 | - | 8 - 9 | 1316 | 120 | ||
| BC052865.1 | - | 8 - 9 | 1212 | 30072864 | 120 | |
More Information
Set Membership: |
|

Cd96
Genomic Map