| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1600013758.mm.1 |
NM_021495.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013758.mm.1 |
NM_021495.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013758.mm.1 |
TC1600001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013759.mm.1 |
NM_021495.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013759.mm.1 |
NM_021495.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013759.mm.1 |
TC1600001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013760.mm.1 |
NM_021495.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013760.mm.1 |
NM_021495.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013760.mm.1 |
TC1600001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013762.mm.1 |
NM_021495.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013762.mm.1 |
NM_021495.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013762.mm.1 |
TC1600001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013763.mm.1 |
NM_021495.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013763.mm.1 |
NM_021495.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013763.mm.1 |
TC1600001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013764.mm.1 |
NM_021495.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013764.mm.1 |
NM_021495.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013764.mm.1 |
NM_021496.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013764.mm.1 |
NM_021496.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013764.mm.1 |
NM_021497.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013764.mm.1 |
NM_021497.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013764.mm.1 |
TC1600001679.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1600013761.mm.1 |
NM_021495.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013761.mm.1 |
NM_021495.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1600013761.mm.1 |
TC1600001679.mm.1 |
MTA TC |