Assay Details
| Gene Symbol: Mast3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199308.2 | NP_955012.2 | 6 - 7 | 685 | 50 | |
| GenBank mRNA | AK155568.1 | - | 7 - 8 | 657 | 50 | |
| CB524773.1 | - | 2 - 3 | 119 | 50 | ||
More Information
Set Membership: |
|

Mast3
Genomic Map