Assay Details
| Gene Symbol: Pde4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201607.2 | NP_963901.1 | 9 - 10 | 970 | 91 | |
| GenBank mRNA | AK149514.1 | - | 9 - 10 | 920 | 91 | |
| BC030873.1 | - | 9 - 10 | 970 | 4223352 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde4c
Genomic Map