Assay Details
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173013.3 | NP_766601.2 | 3 - 4 | 339 | 93 | |
GenBank mRNA | AK032300.1 | - | 3 - 4 | 327 | 93 | |
AK082854.1 | - | 3 - 4 | 333 | 93 | ||
AK158860.1 | - | 1 - 2 | 108 | 93 | ||
BB857804.1 | - | 3 - 4 | 341 | 93 | ||
BC058166.1 | - | 3 - 4 | 320 | 93 | ||
BC150729.1 | - | 3 - 4 | 381 | 9087640 | 93 | |
DQ387862.1 | - | 3 - 4 | 327 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
