Assay Details
| Gene Symbol: Sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133704.4 | NP_598465.2 | 1 - 2 | 95 | 83 | |
| GenBank mRNA | AK007341.1 | - | 1 - 2 | 35 | 83 | |
| AK082516.1 | - | 1 - 2 | 95 | 83 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK082516.1, NM_133704.4 transcripts.
Set Membership: |
|

Sec22a
Genomic Map