Assay Details
| Gene Symbol: Sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133704.4 | NP_598465.2 | 2 - 3 | 296 | 110 | |
| GenBank mRNA | AK007341.1 | - | 2 - 3 | 236 | 110 | |
| AK044615.1 | - | 2 - 3 | 231 | 110 | ||
| AK082516.1 | - | 2 - 3 | 296 | 110 | ||
| BC138969.1 | - | 1 - 2 | 210 | 8861991 | 110 | |
| BC138971.1 | - | 1 - 2 | 210 | 8861993 | 110 | |
| BC145477.1 | - | 1 - 2 | 212 | 110 | ||
| BC145478.1 | - | 1 - 2 | 212 | 9054040 | 110 | |
More Information
Set Membership: |
|

Sec22a
Genomic Map