Assay Details
Gene Symbol: Pros1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011173.2 | NP_035303.1 | 13 - 14 | 1738 | 76 | |
GenBank mRNA | AK133044.1 | - | 2 - 3 | 210 | 76 | |
AK145501.1 | - | 13 - 14 | 1738 | 76 | ||
AK163032.1 | - | 13 - 14 | 1766 | 76 | ||
BC128316.1 | - | 13 - 14 | 1700 | 40109789 | 76 | |
L27439.1 | - | 12 - 13 | 1549 | 76 | ||
Z25469.1 | - | 13 - 14 | 1704 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
