Assay Details
Gene Symbol: Rabl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042499.1 | NP_001035964.1 | 6 - 7 | 629 | 83 | |
GenBank mRNA | AK011280.1 | - | 6 - 7 | 571 | 83 | |
AK016099.1 | - | 6 - 7 | 638 | 83 | ||
AK028195.1 | - | 6 - 7 | 623 | 83 | ||
AK028269.1 | - | 6 - 7 | 616 | 83 | ||
BC050194.1 | - | 6 - 7 | 610 | 30022531 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
