Assay Details
| Gene Symbol: Hcls1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008225.2 | NP_032251.2 | 11 - 12 | 1112 | 86 | |
| GenBank mRNA | AK088723.1 | - | 11 - 12 | 1094 | 86 | |
| AK132608.1 | - | 2 - 3 | 160 | 86 | ||
| BC007469.1 | - | 11 - 12 | 1082 | 3593456 | 86 | |
| CA321698.1 | - | 2 - 3 | 194 | 86 | ||
| D42120.1 | - | 11 - 12 | 1043 | 86 | ||
| X84797.1 | - | 11 - 12 | 1080 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hcls1
Genomic Map